Learn More
TARGETMOL CHEMICALS INC ELB-139 1MG
SDP
Catalog No. 502877404
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. ELB-139 is a GABAA receptor agonist that can be used to study depression and epilepsy. Purity 100%
Catalog No. 50-287-7404 Supplier TARGETMOL CHEMICALS INC Supplier No. T272521MG
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. ELB-139 is a GABAA receptor agonist that can be used to study depression and epilepsy. Purity 100%