Learn More
TARGETMOL CHEMICALS INC ELBASVIR 10MG
SDP
Catalog No. 502576753
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elbasvir (MK-8742) is an inhibitor of nonstructural protein 5A (NS5A) of hepatitis C virus (HCV) being developed as a component of treatment regimens for chronic HCV infection. purity: 99%
Catalog No. 50-257-6753 Supplier TARGETMOL CHEMICALS INC Supplier No. T454410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elbasvir (MK-8742) is an inhibitor of nonstructural protein 5A (NS5A) of hepatitis C virus (HCV) being developed as a component of treatment regimens for chronic HCV infection. purity: 99%