Learn More
Selleck Chemical LLC Elbasvir
SDP
Catalog No. 5000003033
Encompass_Preferred
Elbasvir (MK8742) is an NS5A inhibitor preventing hepatitis C viral RNA replication and virion assembly Median EC50 values range from 0 2 to 3600 pmol/L based on genotype
Catalog No. 50-000-03033 Supplier Selleck Chemical LLC Supplier No. S56522MG
Elbasvir (MK8742) is an NS5A inhibitor preventing hepatitis C viral RNA replication and virion assembly Median EC50 values range from 0 2 to 3600 pmol/L based on genotype