Learn More
AdipoGen Elbasvir (5 mg)
SDP
Supplier:  AdipoGen AGCR13729M005
Encompass_Preferred
Chemical. CAS: 1370468-36-2. Formula: C49H55N9O7. MW: 882. Elbasvir is a direct-acting antiviral drug that potently inhibits hepatitis C virus (HCV) non-structural protein 5A (NS5A). NS5A is essential for HCV RNA synthesis and viral assembly, and elbasvir inhibits the assembly of new viral replicase complexes when used at a concentration of 50 pM. It exhibits potent inhibitory activity against a broad range of HCV genotypes. Formulations containing elbasvir have been used alone and in combination with NS3/4A protease inhibitors in the treatment of HCV. Identified as a potential inhibitor of SARS-CoV-2, responsible for COVID-19.
Catalog No. 50-205-4141