Learn More
AVANTOR MASTERFLEX (VWR LLC) ELBOW 1/8FTFTX1/8FTFT PP 10/PK
Catalog No. 502742169
Encompass
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 1/8IN ID x 1/8IN NPT(M)10/PK, Avantor
Catalog No. 50-274-2169 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4151878
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 1/8IN ID x 1/8IN NPT(M)10/PK, Avantor