Learn More
AVANTOR MASTERFLEX (VWR LLC) ELBOW 1/8IN X5/16IN 100/PK
Catalog No. 502742262
Encompass
Masterflex Fitting, Polypropylene, Elbow, Hosebarb to Thread, 5/16IN ID x 1/8IN NPT(M)100/PK, Avantor
Catalog No. 50-274-2262 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4062686
Masterflex Fitting, Polypropylene, Elbow, Hosebarb to Thread, 5/16IN ID x 1/8IN NPT(M)100/PK, Avantor