Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Elbow, Hose Barb to Threaded Adapter, 1/4"" ID x 1/8"" NPT(M); 10/PK
Catalog No. 50629867
Encompass
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. 50-629-867 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3040104
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).