Learn More
AVANTOR MASTERFLEX (VWR LLC) John Guest Elbow, 3/16"" OD, Acetal, 10/Pk
Catalog No. NC1650258
Encompass
A single fitting offers secure connections and quick disconnection¿no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop¿steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth¿ tubing slides right out. Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon or silicone, order soft tube supports.
Catalog No. NC1650258 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3400631
A single fitting offers secure connections and quick disconnection¿no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop¿steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth¿ tubing slides right out. Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon or silicone, order soft tube supports.