Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, 316L Stainless Steel, 45 Elbow, Sanitary Clamp Union, 3/4
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3070084
Encompass
These 45-degree elbow adapters allow for secure connection of sanitary lines when a 45-degree angle is required. The 316L stainless steel couplings are suitable for the pharmaceutical industry. They offer great chemical compatibility for use with corrosive acids and bases. Clamps and gaskets required for a complete system are sold separately.
Catalog No. NC1481802