Learn More
Heidolph™ Elbow for Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware
SDP
Catalog No. 13900902 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-902 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-902 Supplier Heidolph™ Supplier No. 036210180
Add to Cart
Add to Cart
Elbow connects expansion vessel to condenser.
Included with glassware sets A, AC,A1, A1C, A2, and A2C
Specifications
Type Glassware
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov