Learn More
Ibidi USA Elbow Luer Connector Male, 25 pcs/box
SDP
Catalog No. NC0571367
Encompass
Elbow Luer Connector Male is a male Luer to single barb connector designed for use with pumps and ibidi µ-Slide products. natural polyethylene, sterile, 25 pcs./box
This item is not returnable. View return policy
Catalog No. NC0571367 Supplier Ibidi USA Supplier No. 10802
Elbow Luer Connector Male is a male Luer to single barb connector designed for use with pumps and ibidi µ-Slide products. natural polyethylene, sterile, 25 pcs./box