Learn More
Grainger Solo Elbow Nozzle Assembly, For Sprayers, Includes Adjustable Nozzle, 1 ea
Catalog No. 502785906
Encompass_Preferred
This elbow nozzle assembly is designed to attach to a spray wand for controlled liquid application patterns. It is engineered as a genuine replacement part to maintain the performance and functionality of compatible spraying equipment.
  • Includes a plastic adjustable nozzle
  • Contains an elbow and a filter element
  • Compatible with all sprayers except one-hand sprayers and ECS series models
  • Manufactured in the United States
Catalog No. 50-278-5906 Supplier Grainger Supplier No. 190T37
This elbow nozzle assembly is designed to attach to a spray wand for controlled liquid application patterns. It is engineered as a genuine replacement part to maintain the performance and functionality of compatible spraying equipment.
  • Includes a plastic adjustable nozzle
  • Contains an elbow and a filter element
  • Compatible with all sprayers except one-hand sprayers and ECS series models
  • Manufactured in the United States