Learn More
Nuaire Inc Large Biosafety Cabinet Elbow Rest, 10 in Length x 3 in Width x 2 in Height
SDP
Catalog No. NC9322045
Encompass
Ergonomic Design of the elbow rest keeps you elevated above the front grill so airflows are not disrupted yet keeps your arms and shoulder in a comfortable position to perform your work. Comfortable close-cell sponge padding does not absorb liquids. Conveniently placed on front air grill of a Biological Safety Cabinet. Fits all makes and models of Biological Safety Cabinets. Easy to clean and autoclavable. Rubber feet caps included for use on flat surfaces such as countertops.
Catalog No. NC9322045 Supplier Nuaire Inc Supplier No. NU978002
Ergonomic Design of the elbow rest keeps you elevated above the front grill so airflows are not disrupted yet keeps your arms and shoulder in a comfortable position to perform your work. Comfortable close-cell sponge padding does not absorb liquids. Conveniently placed on front air grill of a Biological Safety Cabinet. Fits all makes and models of Biological Safety Cabinets. Easy to clean and autoclavable. Rubber feet caps included for use on flat surfaces such as countertops.