Learn More
AVANTOR MASTERFLEX (VWR LLC) ELBOWS 1/2FTFT X 3/8FTFT 10/P
Catalog No. 502742348
Encompass
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 3/8IN ID x 1/2IN NPT(M)10/PK, Avantor
Catalog No. 50-274-2348 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061063
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 3/8IN ID x 1/2IN NPT(M)10/PK, Avantor