Learn More
AVANTOR MASTERFLEX (VWR LLC) ELBOWS 3/4IN X 1/2IN 50/PK
Catalog No. 502742208
Encompass
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 1/2IN ID x 3/4IN NPT(M)50/PK, Avantor
Catalog No. 50-274-2208 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4064068
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 1/2IN ID x 3/4IN NPT(M)50/PK, Avantor