Learn More
AVANTOR MASTERFLEX (VWR LLC) ELBOWS 3/4IN X 3/8IN 5/PK
Catalog No. 502742344
Encompass
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 3/8IN ID x 3/4IN NPT(M)5/PK, Avantor
Catalog No. 50-274-2344 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061067
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 3/8IN ID x 3/4IN NPT(M)5/PK, Avantor