Learn More
AVANTOR MASTERFLEX (VWR LLC) ELBOWS 3/8FTFT X 1/4FTFT 10/P
Catalog No. 502742352
Encompass
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 1/4IN ID x 3/8IN NPT(M)10/PK, Avantor
Catalog No. 50-274-2352 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061059
Masterflex Fitting, Natural Polypropylene, Elbow, Hosebarb to Thread, 1/4IN ID x 3/8IN NPT(M)10/PK, Avantor