Learn More
NATUS MEDICAL INC Disposable 2x2 Adhesive Electrode Set with Lead Wires Attached, 59” cable length
Catalog No. NC1691236
Encompass
Color-coded (two red, two black) attached lead wires terminate in 1.5 mm female touchproof connectors. Four pre-gelled 20 mm Ag/AgCl disc electrodes per set. Ideal for IOM applications.
This item is not returnable. View return policy
Catalog No. NC1691236 Supplier NATUS MEDICAL INC Supplier No. 019415000
Color-coded (two red, two black) attached lead wires terminate in 1.5 mm female touchproof connectors. Four pre-gelled 20 mm Ag/AgCl disc electrodes per set. Ideal for IOM applications.