Learn More
CPC Scientific H-Ser-Asn-Leu-Ser-Thr-Asu-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asp-Val-Gly-Ala-Gly-Thr-Pro-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OSTP013B
Encompass
Carbocalcitonin, the amino-suberic acid analog of eel calcitonin, has all the biological properties of the corresponding natural calcitonin. The substitution of the disulfide bond of natural calcitonins with an ethylene bridge in the 1-7 N-terminal position gives elcatonin greater stability and excellent tolerability when used in vivo. ONE-LETTER SEQUENCE: SNLST-Asu-VLGKLSQELHKLQTYPRTDVGAGTP-NH2
(Peptide bond Ser1 and Asu6 bridge)MOLECULAR FORMULA: C147H242N42O47MOLECULAR WEIGHT:3363.82STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [60731-46-6], SYNONYMS: Elcatonin, (Asu)-Calcitonin (eel), (Des-Cys,cyclo(Ser-Asu))-Calcitonin (eel)RESEARCH AREA: OsteoporosisREFERENCES: A. Caniggia et al., Minerva Medica , 74, 993 (1983); Italian Osteoporosis Network, Current Therapeutic Res., 45, 502 (1989)
Catalog No. 50-211-0022