Learn More
MSC Elcometer Cross Hatch Cutter, Adhesion Testing, 4-Sided Cutter
Catalog No. 5000516898
Encompass
This cross hatch cutter is designed for adhesion tests, offering an immediate assessment of the bond quality to the substrate. Its robust construction makes it suitable for thin, thick, or tough coatings across various surfaces, serving as an effective tool for both field and laboratory use.
  • Robust design
  • Large, non-slip grip
  • Ideal for thin, thick, or hard coatings
  • Quick-change, four-sided cutter for adhesion testing across a range of coating thicknesses (1 mm, 1.5 mm, 2 mm, and 3 mm)
Catalog No. 50-005-16898 Supplier MSC Supplier No. 06117493
This cross hatch cutter is designed for adhesion tests, offering an immediate assessment of the bond quality to the substrate. Its robust construction makes it suitable for thin, thick, or tough coatings across various surfaces, serving as an effective tool for both field and laboratory use.
  • Robust design
  • Large, non-slip grip
  • Ideal for thin, thick, or hard coatings
  • Quick-change, four-sided cutter for adhesion testing across a range of coating thicknesses (1 mm, 1.5 mm, 2 mm, and 3 mm)