Learn More
MSC Elcometer USB Cable, USB to Mini-USB, 2 m
Catalog No. 5000529328
Encompass
This USB cable is designed to facilitate data transfer between instruments, such as the Elcometer 406, and a computer. It features a USB to Mini-USB connection and has a length of 2 meters (approximately 6.5 feet), providing ample reach for connecting devices.
  • Facilitates data transfer between instruments and computers.
  • Features a USB to Mini-USB connector type.
  • Provides a length of 2 meters (6.5 ft).
  • Compatible with various instruments.
Catalog No. 50-005-29328 Supplier MSC Supplier No. 06118053
This USB cable is designed to facilitate data transfer between instruments, such as the Elcometer 406, and a computer. It features a USB to Mini-USB connection and has a length of 2 meters (approximately 6.5 feet), providing ample reach for connecting devices.
  • Facilitates data transfer between instruments and computers.
  • Features a USB to Mini-USB connector type.
  • Provides a length of 2 meters (6.5 ft).
  • Compatible with various instruments.