Learn More
Grainger Elct Flr Htr Thrmostat 4-3/4 H 1 Pole
Catalog No. 502753559
Encompass_Preferred
The Dayton Electric Floor Heater Thermostat is a replacement thermostat for Dayton electric floor heaters. It has a dial for temperature adjustment from 40 to 90 degrees Fahrenheit. The thermostat measures 4-3/4 inches wide x 4-3/4 inches high x 2-1/2 inches deep. It is made of plastic with a white finish and mounts into a standard junction box. The thermostat requires a 120V power connection and controls a resistive load up to 15 amps. It can be used as a replacement part for Dayton electric floor heaters.
Catalog No. 50-275-3559 Supplier Grainger Supplier No. 14C466
The Dayton Electric Floor Heater Thermostat is a replacement thermostat for Dayton electric floor heaters. It has a dial for temperature adjustment from 40 to 90 degrees Fahrenheit. The thermostat measures 4-3/4 inches wide x 4-3/4 inches high x 2-1/2 inches deep. It is made of plastic with a white finish and mounts into a standard junction box. The thermostat requires a 120V power connection and controls a resistive load up to 15 amps. It can be used as a replacement part for Dayton electric floor heaters.