Learn More
Electron Microscopy Sciences Lab Aire Electric Bench Top Dryer Single Side 3 Tier
SDP
Catalog No. 501902089
Encompass_Preferred
Electric benchtop dryers offer a compact footprint and accelerate drying of labware with 400 watts of gently circulated heated air. Heat selector knob with three temperature settings: high, medium and low. Power button illuminates when in use, and an 8-amp @ 120V (4-amp @ 230V) circuit breaker and independent thermo-fuse protects unit from overheating. Electric drying efficiency is enhanced with a replaceable polyurethane foam filter that keeps warmed air dust free. Removable polypropylene baskets serve as storage for small labware and extra pegs.
Includes: 3 Tier (2 Baskets) with 38 pegs
Catalog No. 50-190-2089 Supplier Electron Microscopy Sciences Supplier No. 6440003
Electric benchtop dryers offer a compact footprint and accelerate drying of labware with 400 watts of gently circulated heated air. Heat selector knob with three temperature settings: high, medium and low. Power button illuminates when in use, and an 8-amp @ 120V (4-amp @ 230V) circuit breaker and independent thermo-fuse protects unit from overheating. Electric drying efficiency is enhanced with a replaceable polyurethane foam filter that keeps warmed air dust free. Removable polypropylene baskets serve as storage for small labware and extra pegs.
Includes: 3 Tier (2 Baskets) with 38 pegs