Learn More
Medchemexpress LLC Eldecalcitol | 104121-92-8 | 99.9% | C30H50O5 | 5 MG
SDP
Catalog No. 5000433533
Encompass_Preferred
Eldecalcitol (ED-71) is an orally active vitamin D3 analogue that inhibits bone resorption and increases bone mineral density. It also exhibits an anti-tumor effect, inhibits cell proliferation, migration, and induces apoptosis by suppressing GPx-1.
  • Orally active vitamin D3 analogue
  • Inhibits bone resorption and increases bone mineral density
  • Exhibits anti-tumor effects
  • Inhibits cell proliferation and migration
  • Induces apoptosis by suppressing GPx-1
  • Shows no cytotoxicity at 0-50 nM for 24 hours
  • Reduces cell death induced by LPS
  • Inhibits LPS-induced pyroptosis by activating Nrf2 and its effector molecule HO-1
  • Arrests cell cycle at G0/G1 phase
  • Stimulates focal bone formation and suppresses bone resorption
Catalog No. 50-004-33533 Supplier Medchemexpress LLC Supplier No. HYA00205MG
Eldecalcitol (ED-71) is an orally active vitamin D3 analogue that inhibits bone resorption and increases bone mineral density. It also exhibits an anti-tumor effect, inhibits cell proliferation, migration, and induces apoptosis by suppressing GPx-1.
  • Orally active vitamin D3 analogue
  • Inhibits bone resorption and increases bone mineral density
  • Exhibits anti-tumor effects
  • Inhibits cell proliferation and migration
  • Induces apoptosis by suppressing GPx-1
  • Shows no cytotoxicity at 0-50 nM for 24 hours
  • Reduces cell death induced by LPS
  • Inhibits LPS-induced pyroptosis by activating Nrf2 and its effector molecule HO-1
  • Arrests cell cycle at G0/G1 phase
  • Stimulates focal bone formation and suppresses bone resorption