Learn More
Medchemexpress LLC Eldecalcitol-d6 | 104121-92-8 | 99.5% | C30H44D6O5 | 1 MG
SDP
Catalog No. 501932893
Encompass_Preferred
Eldecalcitol-d6 is the deuterium-labeled Eldecalcitol, an orally active analogue of active vitamin D used in the treatment of osteoporosis. It has a strong inhibitory effect on bone resorption and causes a significant increase in bone mineral density.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-193-2893 Supplier Medchemexpress LLC Supplier No. HYA0020S1MG
Eldecalcitol-d6 is the deuterium-labeled Eldecalcitol, an orally active analogue of active vitamin D used in the treatment of osteoporosis. It has a strong inhibitory effect on bone resorption and causes a significant increase in bone mineral density.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS