Learn More
Protexer Corporation Electric Shoe Cover Refill - CPE - 1,680 pairs/ case
Catalog No. NC2228095
Encompass_Preferred
For use with Electric BootieButler Dispensers (EMLG) only. Impervious shoe cover Constructed of Chlorinated Polyethylene material. One size fits most (up to men’s size 15). Approved for Class 10,000/ISO-7 Cleanrooms.
This item is not returnable. View return policy
Catalog No. NC2228095 Supplier Protexer Corporation Supplier No. EB1680CPE
For use with Electric BootieButler Dispensers (EMLG) only. Impervious shoe cover Constructed of Chlorinated Polyethylene material. One size fits most (up to men’s size 15). Approved for Class 10,000/ISO-7 Cleanrooms.