Learn More
Medchemexpress LLC Elebsiran sodium (VIR-2218 sodium) | 00-00-0 | 95.8% | C485H658F9N158O294P39S6 | 10MG
SDP
Catalog No. 5000258226
Encompass_Preferred
Elebsiran sodium is a research-grade small interfering RNA (siRNA) sodium salt designed to target and degrade hepatitis B virus (HBV) and hepatitis D virus (HDV) RNA transcripts. It is GalNAc-conjugated for liver-targeted delivery and is intended for laboratory research into antiviral activity and antigen reduction.
  • Targets HBV and HDV RNA transcripts for RNAi-mediated degradation.
  • GalNAc conjugation enables liver-targeted delivery via ASGPR.
  • Demonstrates reduction of HBsAg and viral load in studies.
  • Provided as the sodium salt suitable for in vitro and in vivo research.
  • High purity appropriate for biological assays (95.8%).
Catalog No. 50-002-58226 Supplier Medchemexpress LLC Supplier No. HY14726610MG
Elebsiran sodium is a research-grade small interfering RNA (siRNA) sodium salt designed to target and degrade hepatitis B virus (HBV) and hepatitis D virus (HDV) RNA transcripts. It is GalNAc-conjugated for liver-targeted delivery and is intended for laboratory research into antiviral activity and antigen reduction.
  • Targets HBV and HDV RNA transcripts for RNAi-mediated degradation.
  • GalNAc conjugation enables liver-targeted delivery via ASGPR.
  • Demonstrates reduction of HBsAg and viral load in studies.
  • Provided as the sodium salt suitable for in vitro and in vivo research.
  • High purity appropriate for biological assays (95.8%).