Learn More
MSC PRO-SOURCE Pressure Washer, 3.0 GPM, 2500 PSI, Electric, Cold Water
Catalog No. 502837615
Encompass
The electric cold water pressure washer is designed for demanding cleaning tasks, delivering a powerful 2500 PSI and a flow rate of 3.0 GPM. It features a durable AR Triplex Ceramic Plunger Pump for reliable performance.
  • Designed for cold water applications.
  • Electric powered for consistent and reliable operation.
  • Delivers a flow rate of 3.0 GPM.
  • Operates at 230 voltage.
  • Maximum amperage of 25.0 A.
  • Features an AR Triplex Ceramic Plunger pump.
  • Single power phase for compatibility.
  • Includes a 50.00 foot hose.
  • Equipped with a 2-wheel cart frame for portability.
  • Provides 2500 PSI of pressure.
  • Hose size is 3/8 "
Catalog No. 50-283-7615 Supplier MSC Supplier No. 48352322
Add to Cart
Add to Cart
Request a Quote
The electric cold water pressure washer is designed for demanding cleaning tasks, delivering a powerful 2500 PSI and a flow rate of 3.0 GPM. It features a durable AR Triplex Ceramic Plunger Pump for reliable performance.
  • Designed for cold water applications.
  • Electric powered for consistent and reliable operation.
  • Delivers a flow rate of 3.0 GPM.
  • Operates at 230 voltage.
  • Maximum amperage of 25.0 A.
  • Features an AR Triplex Ceramic Plunger pump.
  • Single power phase for compatibility.
  • Includes a 50.00 foot hose.
  • Equipped with a 2-wheel cart frame for portability.
  • Provides 2500 PSI of pressure.
  • Hose size is 3/8 "