Learn More
MSC Value Collection Pressure Washer, Cold Water, Electric, 3.0 GPM, 220 V
Catalog No. 502838131
Encompass
Value Collection pressure washers are designed for effective cleaning with cold water. This electric model features a 3.0 GPM flow rate and operates on 220V, making it suitable for various industrial and commercial applications.
  • Cold water type
  • Electric power type
  • 3.0 GPM flow rate
  • 220 V voltage
  • 20.0 A maximum amperage
  • AR Triplex Ceramic Plunger pump type
  • Single power phase
  • 50.00 ft hose length
  • 2000 PSI pressure
  • 3/8 in hose size
  • 2 wheel cart frame
Catalog No. 50-283-8131 Supplier MSC Supplier No. 48352330
Value Collection pressure washers are designed for effective cleaning with cold water. This electric model features a 3.0 GPM flow rate and operates on 220V, making it suitable for various industrial and commercial applications.
  • Cold water type
  • Electric power type
  • 3.0 GPM flow rate
  • 220 V voltage
  • 20.0 A maximum amperage
  • AR Triplex Ceramic Plunger pump type
  • Single power phase
  • 50.00 ft hose length
  • 2000 PSI pressure
  • 3/8 in hose size
  • 2 wheel cart frame