Learn More
MSC Karcher Pressure Washer, 1.8 GPM, 1300 PSI, 120V, 1 ea
Catalog No. 502817743
Encompass
The HD 1.8/13 C is a compact cold water pressure washer designed for maximum portability and maneuverability. It can be stored, transported, and operated in both vertical and horizontal positions. Weighing 55.6 lbs, this professional unit is easily moved by one person.
  • Compact design with integrated carrying and retractable push handle for easy transport
  • Flexible operation in vertical or horizontal positions for maximum stability
  • EASY!Force high-pressure gun reduces user fatigue
  • EASY!Lock quick-release couplings are durable, robust, and five times faster than traditional screw connections
  • Automatic pressure relief and high-quality brass cylinder head extend service life
  • Large, easy-to-reach water fine filter protects the pump from dirt particles
  • Integrated accessory storage for surface cleaner, nozzles, and pressure hose
  • Backed by a 7-year pump warranty
Catalog No. 50-281-7743 Supplier MSC Supplier No. 16616815
The HD 1.8/13 C is a compact cold water pressure washer designed for maximum portability and maneuverability. It can be stored, transported, and operated in both vertical and horizontal positions. Weighing 55.6 lbs, this professional unit is easily moved by one person.
  • Compact design with integrated carrying and retractable push handle for easy transport
  • Flexible operation in vertical or horizontal positions for maximum stability
  • EASY!Force high-pressure gun reduces user fatigue
  • EASY!Lock quick-release couplings are durable, robust, and five times faster than traditional screw connections
  • Automatic pressure relief and high-quality brass cylinder head extend service life
  • Large, easy-to-reach water fine filter protects the pump from dirt particles
  • Integrated accessory storage for surface cleaner, nozzles, and pressure hose
  • Backed by a 7-year pump warranty