Learn More
MSC Value Collection Pressure Washer, Hot Water, Electric, 4.0 GPM, 2300 PSI
Catalog No. 502836610
Encompass
This Value Collection pressure washer is an electric, hot water unit designed for heavy-duty cleaning tasks. It features a General Triplex Ceramic Plunger pump, providing a flow rate of 4.0 GPM and a maximum pressure of 2300 PSI.
  • Quality products at a great value to support your business needs.
  • Hot water type for effective cleaning.
  • Electric power for reliable operation.
  • 4.0 GPM flow rate for efficient cleaning.
  • 2300 PSI pressure for demanding applications.
  • General Triplex Ceramic Plunger pump for durability.
  • 50 foot hose included for extended reach.
  • 4 wheel frame for easy maneuverability.
  • Made in the United States.
Catalog No. 50-283-6610 Supplier MSC Supplier No. 48352678
Add to Cart
Add to Cart
Request a Quote
This Value Collection pressure washer is an electric, hot water unit designed for heavy-duty cleaning tasks. It features a General Triplex Ceramic Plunger pump, providing a flow rate of 4.0 GPM and a maximum pressure of 2300 PSI.
  • Quality products at a great value to support your business needs.
  • Hot water type for effective cleaning.
  • Electric power for reliable operation.
  • 4.0 GPM flow rate for efficient cleaning.
  • 2300 PSI pressure for demanding applications.
  • General Triplex Ceramic Plunger pump for durability.
  • 50 foot hose included for extended reach.
  • 4 wheel frame for easy maneuverability.
  • Made in the United States.