Learn More
MSC Value Collection Pressure Washer, Electric, Hot Water, 3.0 GPM, Triplex Plunger Pump
Catalog No. 502838512
Encompass
This electric hot water pressure washer provides a robust 3.0 GPM flow rate for effective cleaning. Featuring a durable triplex plunger pump and 2400 PSI pressure, it is suitable for demanding applications, operating at 220 V with a maximum amperage of 25.0 A.
  • Electric hot water operation
  • 3.0 GPM flow rate
  • Triplex plunger pump
  • 220 V voltage
  • 25.0 A maximum amperage
  • 50 ft. hose length
  • 2400 PSI pressure
  • 4 wheel box frame
Catalog No. 50-283-8512 Supplier MSC Supplier No. 48352660
Add to Cart
Add to Cart
Request a Quote
This electric hot water pressure washer provides a robust 3.0 GPM flow rate for effective cleaning. Featuring a durable triplex plunger pump and 2400 PSI pressure, it is suitable for demanding applications, operating at 220 V with a maximum amperage of 25.0 A.
  • Electric hot water operation
  • 3.0 GPM flow rate
  • Triplex plunger pump
  • 220 V voltage
  • 25.0 A maximum amperage
  • 50 ft. hose length
  • 2400 PSI pressure
  • 4 wheel box frame