Learn More
METTLER TOLEDO pH Sensor InPro 3250i SG/425
Catalog No. NC0270052 Shop All METTLER TOLEDO Products
Encompass
The InPro 3250i SG/425 is a pre-pressurized, liquid filled, low maintenance pH & ORP sensor, combined with a temperature sensor, for in-line measurement in demanding applications. The 425 mm version is optimal for long installation lengths.
Catalog No. NC0270052 Supplier METTLER TOLEDO Supplier No. 52005376
The InPro 3250i SG/425 is a pre-pressurized, liquid filled, low maintenance pH & ORP sensor, combined with a temperature sensor, for in-line measurement in demanding applications. The 425 mm version is optimal for long installation lengths.