Learn More
MSC SPI Swiss Precision Instruments Electronic Inside Micrometer Set, Carbide Measuring Face, 80 mm Gage Depth, 1 ea
Catalog No. 5000513424
Encompass
These internal micrometer sets are designed for precision measurements, including capabilities for blind hole gauging. Each unit features a calibrated system with a rotating spindle and a ratchet stop thimble to ensure consistent contact pressure. The measuring contact points are specifically engineered to reach near the bottom of blind holes.
  • Measuring contact points designed for blind hole measurements
  • Includes extension rods for increased depth reach up to 6 in.
  • Equipped with a ratchet stop thimble for consistent pressure
  • Calibrated to ASME 89.1.13 standards
  • Features a digital data output port
  • Includes one 3V CR2032 lithium battery
  • Carbide measuring faces for durability
Catalog No. 50-005-13424 Supplier MSC Supplier No. 13433578
These internal micrometer sets are designed for precision measurements, including capabilities for blind hole gauging. Each unit features a calibrated system with a rotating spindle and a ratchet stop thimble to ensure consistent contact pressure. The measuring contact points are specifically engineered to reach near the bottom of blind holes.
  • Measuring contact points designed for blind hole measurements
  • Includes extension rods for increased depth reach up to 6 in.
  • Equipped with a ratchet stop thimble for consistent pressure
  • Calibrated to ASME 89.1.13 standards
  • Features a digital data output port
  • Includes one 3V CR2032 lithium battery
  • Carbide measuring faces for durability