Learn More
MSC Buyers Products Electric Poly Hopper Spreader, 2 cu. yd., 12V DC, Pintle Chain
Catalog No. 502840016
Encompass
This electric poly hopper spreader features a 2 cubic yard capacity and a 12V DC-powered electric drive with an independent pintle chain conveyor. Designed for 3/4 and 1 ton pickup trucks with 6-1/2 ft beds and minimum 10,000 lb GVW, it effectively handles bulk salt, sand, or a 50/50 sand/salt mix. The durable polyethylene hopper ensures longevity and resistance to rust or dents.
  • Independent conveyor and spinner motor controls
  • 30 ft maximum spread width
  • Durable, patented dual-wall poly hopper
  • In-cab controls with independent speed settings
  • 12 inch all-steel pintle chain conveyor
  • 14 inch poly spinner
  • Swing-away chute for easy cleaning and bulk unloading
  • Vibrator, top screen, fitted tarp, and tie downs included
  • Forklift slots for easy loading
  • Made in the USA
  • Two year warranty
Catalog No. 50-284-0016 Supplier MSC Supplier No. 94790409
Add to Cart
Add to Cart
Request a Quote
This electric poly hopper spreader features a 2 cubic yard capacity and a 12V DC-powered electric drive with an independent pintle chain conveyor. Designed for 3/4 and 1 ton pickup trucks with 6-1/2 ft beds and minimum 10,000 lb GVW, it effectively handles bulk salt, sand, or a 50/50 sand/salt mix. The durable polyethylene hopper ensures longevity and resistance to rust or dents.
  • Independent conveyor and spinner motor controls
  • 30 ft maximum spread width
  • Durable, patented dual-wall poly hopper
  • In-cab controls with independent speed settings
  • 12 inch all-steel pintle chain conveyor
  • 14 inch poly spinner
  • Swing-away chute for easy cleaning and bulk unloading
  • Vibrator, top screen, fitted tarp, and tie downs included
  • Forklift slots for easy loading
  • Made in the USA
  • Two year warranty