Learn More
MSC Buyers Products Electric Poly Hopper Spreader, Auger, 2 cubic yard, 12V DC
Supplier: MSC 94790722
This electric poly hopper spreader is designed for commercial and residential use, spreading bulk salt or a 50/50 sand/salt mix. It features a durable 2 cubic yard dual-wall poly hopper, a 5-1/4 in. steel auger, and a 14 in. poly spinner with up to a 30 ft spread. The 12V DC-powered unit has independent auger and spinner motors, in-cab controls, and fits 3/4 ton and 1 ton pickup trucks.
- Durable, patented dual-wall poly hopper.
- 12V DC-powered electric drive with independent auger and spinner motors.
- Handles bulk salt or a 50/50 sand/salt mix.
- In-cab controls for independent auger and spinner speeds, with vibrator control.
- Swing-away chute for easy cleaning and storage.
- Includes vibrator, top screen, fitted tarp, tie downs, and remote auger bearing grease kit.
- Integrated forklift slots for easy loading.
- Backed by a 2 year warranty.
- Made in the USA.