Learn More
Protexer Corporation Electric Shoe Cover Refill - Non Woven 1,050 pairs/ case
Catalog No. NC2228105
Encompass_Preferred
For use with Electric BootieButler Dispensers (EMLG) only. Fluid Resistant, Non-Woven shoe cover. One size fits most (up to men’s size 15). Approved for Class 10,000/ISO-7 Cleanrooms.
This item is not returnable. View return policy
Catalog No. NC2228105 Supplier Protexer Corporation Supplier No. EB1050NW
For use with Electric BootieButler Dispensers (EMLG) only. Fluid Resistant, Non-Woven shoe cover. One size fits most (up to men’s size 15). Approved for Class 10,000/ISO-7 Cleanrooms.