Learn More
Grainger Elec Tape 15 ft Lx1 in W 3.5 mil CLR
Supplier:  Grainger 15D549
Encompass_Preferred
The TapeCase Insulating Electrical Tape is a high-quality PVC tape made in the USA. It is 7 mil thick with an operating temperature up to 105°C. It meets UL510 and MIL-I-24391 standards. Available in black, white, red, yellow, and blue, the tape stretches to conform, providing excellent protection. It bonds instantly, resists abrasion, moisture, chemicals, and weather. Ideal for insulating wire and cable splices, terminations, and repairs.
Catalog No. 50-275-0166