Learn More
Grainger Approved Vendor Insulating Electrical Tape, Vinyl, 7 mil, 600 V, 1 ea
Catalog No. 502782891
Encompass_Preferred
These continuous rolls of premium vinyl electrical tape are designed to insulate wire and cable, helping to prevent electrical shorts and shock in general purpose applications. Suitable for both indoor and outdoor use, this tape features a durable rubber adhesive on a vinyl backing.
  • Provides reliable insulation with a voltage rating up to 600 V
  • Withstands a performance temperature range of 30°F to 220°F
  • Resistant to abrasion, chemicals, flame, UV radiation, moisture, and weathering
  • Single-sided adhesive format with a 15 lb./in. tensile strength
  • Meets professional standards including CSA and UL certification
Catalog No. 50-278-2891 Supplier Grainger Supplier No. 19N741
These continuous rolls of premium vinyl electrical tape are designed to insulate wire and cable, helping to prevent electrical shorts and shock in general purpose applications. Suitable for both indoor and outdoor use, this tape features a durable rubber adhesive on a vinyl backing.
  • Provides reliable insulation with a voltage rating up to 600 V
  • Withstands a performance temperature range of 30°F to 220°F
  • Resistant to abrasion, chemicals, flame, UV radiation, moisture, and weathering
  • Single-sided adhesive format with a 15 lb./in. tensile strength
  • Meets professional standards including CSA and UL certification