Learn More
Grainger Approved Vendor Insulating Electrical Tape, Vinyl, 3/4 in, 7 mil, 22 yd, 1 ea
Catalog No. 502779211
Encompass_Preferred
This premium insulating electrical tape is designed for general purpose wire and cable insulation to effectively prevent shorts and electrical shock. Constructed with a durable vinyl backing and a rubber adhesive, this single-sided, continuous roll tape is suitable for both indoor and outdoor applications.
  • 600 V voltage rating
  • 7 mil thickness
  • 15 lb/in. tensile strength
  • 16 oz/in. adhesion to steel
  • Temperature performance range of 32°F to 221°F
  • Abrasion, chemical, flame, moisture, UV, and weather resistant
  • CSA and UL listed
Catalog No. 50-277-9211 Supplier Grainger Supplier No. 19N742
This premium insulating electrical tape is designed for general purpose wire and cable insulation to effectively prevent shorts and electrical shock. Constructed with a durable vinyl backing and a rubber adhesive, this single-sided, continuous roll tape is suitable for both indoor and outdoor applications.
  • 600 V voltage rating
  • 7 mil thickness
  • 15 lb/in. tensile strength
  • 16 oz/in. adhesion to steel
  • Temperature performance range of 32°F to 221°F
  • Abrasion, chemical, flame, moisture, UV, and weather resistant
  • CSA and UL listed