Learn More
Medchemexpress LLC Eleclazine hydrochloride | 1448754-43-5 | 99.7% | 451.83 | 1 ML
SDP
Catalog No. 5000390860
Encompass_Preferred
Eleclazine (GS 6615) hydrochloride is a selective cardiac late sodium current inhibitor and a weak inhibitor of potassium current with IC50 value of <1 μM and approximately 14.2 μM, respectively. Eleclazine hydrochloride shows concurrent protection against autonomically induced atrial premature beats, repolarization alternans and heterogeneity, and atrial fibrillation in porcine model. It can be used to research cardiac arrhythmias.
  • Selective cardiac late sodium current inhibitor
  • Weak inhibitor of potassium current with IC50 values of <1 μM and approximately 14.2 μM
  • Provides protection against autonomically induced atrial premature beats, repolarization alternans and heterogeneity, and atrial fibrillation in a porcine model
  • Can be used for research on cardiac arrhythmias
Catalog No. 50-003-90860 Supplier Medchemexpress LLC Supplier No. HY16738A10MM/1ML
Eleclazine (GS 6615) hydrochloride is a selective cardiac late sodium current inhibitor and a weak inhibitor of potassium current with IC50 value of <1 μM and approximately 14.2 μM, respectively. Eleclazine hydrochloride shows concurrent protection against autonomically induced atrial premature beats, repolarization alternans and heterogeneity, and atrial fibrillation in porcine model. It can be used to research cardiac arrhythmias.
  • Selective cardiac late sodium current inhibitor
  • Weak inhibitor of potassium current with IC50 values of <1 μM and approximately 14.2 μM
  • Provides protection against autonomically induced atrial premature beats, repolarization alternans and heterogeneity, and atrial fibrillation in a porcine model
  • Can be used for research on cardiac arrhythmias