Learn More
Hach Company ELECT.COND.SNSR-CONV-Peek
Catalog No. 5000207324 Shop All Hach Company Products
Encompass
Electrodeless Conductivity Sensor-PEEK-Convertible Style-20 ft Cable-Paper TagPEEK Convertible Mounting Style 6m (20ft) Analog Cable Paper Tag Electrodeless Conductivity Sensor
Catalog No. 50-002-07324 Supplier Hach Company Supplier No. 3727E2T20P
Electrodeless Conductivity Sensor-PEEK-Convertible Style-20 ft Cable-Paper TagPEEK Convertible Mounting Style 6m (20ft) Analog Cable Paper Tag Electrodeless Conductivity Sensor