Learn More
Hach Company ELECT.COND.SNSR-CONV-PFA
Catalog No. 5000207684 Shop All Hach Company Products
Encompass
Electrodeless Conductivity Sensor-Convertible Style-PFA Body MaterialPFA Convertible Mounting Style 6m (20 ft) Analog Cable Generic ID Electrodeless Conductivity Sensor
Catalog No. 50-002-07684 Supplier Hach Company Supplier No. 3728E2T20G
Electrodeless Conductivity Sensor-Convertible Style-PFA Body MaterialPFA Convertible Mounting Style 6m (20 ft) Analog Cable Generic ID Electrodeless Conductivity Sensor