Learn More
Hach Company ELECT.COND.SNSR-CONV-PFA
Catalog No. 5000207732 Shop All Hach Company Products
Encompass
Electrodeless Conductivity Sensor-Convertible Style-PEEK Body Material-50-ft CablePEEK Convertible Mounting Style 15m (50ft) Analog Cable Stainless Steel Tag Electrodeless Conductivity Sensor
Catalog No. 50-002-07732 Supplier Hach Company Supplier No. 3727E2T50N
Electrodeless Conductivity Sensor-Convertible Style-PEEK Body Material-50-ft CablePEEK Convertible Mounting Style 15m (50ft) Analog Cable Stainless Steel Tag Electrodeless Conductivity Sensor