Learn More
Hach Company Filling Solution, Internal, Polarographic DO, 50 mL
Catalog No. 501996638 Shop All Hach Company Products
Encompass
Filling solution for Polarographic Dissolved Oxygen probe, Potassium Chloride/Glycerine, 50 mL bottle
Catalog No. 50-199-6638 Supplier Hach Company Supplier No. 2759126
Filling solution for Polarographic Dissolved Oxygen probe, Potassium Chloride/Glycerine, 50 mL bottle