Learn More
Grainger ELECT TAPE SCOTCH 3M Y 3/4 IN
Catalog No. NC9081127
Encompass_Preferred
Scotch 35 Vinyl Electrical Tape, 3/4, Yellow, Tape is 7 mil thick to provide excellent mechanical and electrical insulation, It wraps smoothly and holds tightly over a wide range of Temperatures, up to 220 deg.F (105 deg.C), 1
Catalog No. NC9081127 Supplier Grainger Supplier No. 2A232
Scotch 35 Vinyl Electrical Tape, 3/4, Yellow, Tape is 7 mil thick to provide excellent mechanical and electrical insulation, It wraps smoothly and holds tightly over a wide range of Temperatures, up to 220 deg.F (105 deg.C), 1