Learn More
HANNA INSTRUMENTS INC 3.5M KCl electrolyte fill solution for double-junction electrodes, 4 x 30 mL
SDP
Catalog No. NC2045111
Encompass
HI7082 is a pH and ORP electrode fill solution that contains 3.5M KCl, which is specifically produced for double junction electrodes. The electrolyte level in refillable electrodes should be checked before performing any measurements. If the level is low, refill with the proper electrolyte solution to ensure optimum performance. This simple maintenance helps guarantee adequate head pressure to promote the flow of reference electrolyte into the sample being measured.
Catalog No. NC2045111 Supplier HANNA INSTRUMENTS INC Supplier No. HI7082
HI7082 is a pH and ORP electrode fill solution that contains 3.5M KCl, which is specifically produced for double junction electrodes. The electrolyte level in refillable electrodes should be checked before performing any measurements. If the level is low, refill with the proper electrolyte solution to ensure optimum performance. This simple maintenance helps guarantee adequate head pressure to promote the flow of reference electrolyte into the sample being measured.