Learn More
MSC MetroVac Electric Sweeper, .75 Peak HP, 120V, All Steel Construction
Catalog No. 502838556
Encompass
This ElectraSweep electric sweeper provides compact and powerful cleaning for various environments. Featuring durable all-steel construction, it delivers reliable performance with a .75 peak horsepower motor operating at 120 V. It offers strong suction with a 60 in water lift sealed suction measurement and is designed for dry pick up only. Essential accessories are included for versatile cleaning tasks.
  • All steel construction
  • .75 Peak HP motor
  • Operates at 120 V, 4 Amps, 500 Watts, 50 CFM
  • Lightweight at 5.5 lbs
  • Includes upholstery nozzle, crevice tool, swivel floor brush, and dust brush
  • 30 ft. 2-conductor cord with quick-release storage strap
  • 60 in water lift sealed suction
  • Designed for dry pick-up only
  • Ergonomic chrome/steel handle
Catalog No. 50-283-8556 Supplier MSC Supplier No. 38775375
This ElectraSweep electric sweeper provides compact and powerful cleaning for various environments. Featuring durable all-steel construction, it delivers reliable performance with a .75 peak horsepower motor operating at 120 V. It offers strong suction with a 60 in water lift sealed suction measurement and is designed for dry pick up only. Essential accessories are included for versatile cleaning tasks.
  • All steel construction
  • .75 Peak HP motor
  • Operates at 120 V, 4 Amps, 500 Watts, 50 CFM
  • Lightweight at 5.5 lbs
  • Includes upholstery nozzle, crevice tool, swivel floor brush, and dust brush
  • 30 ft. 2-conductor cord with quick-release storage strap
  • 60 in water lift sealed suction
  • Designed for dry pick-up only
  • Ergonomic chrome/steel handle