Learn More
Grainger Schneider Electric Electric Actuator, 240V, 2-Position, Aluminum, 60 lb-in.
Catalog No. 502762200
Encompass_Preferred
This 240V electric actuator provides two-position operation for dampers, valves, and related equipment requiring a return to the normal position upon power interruption. The assembly features a rugged, die-cast aluminum enclosure designed for durability and is suitable for use in hazardous locations when configured appropriately. It operates with a two-wire control circuit and utilizes an oil-immersed motor and gear train for reliable performance.
  • 60 lb-in. rated torque
  • Includes auxiliary SPDT switch
  • Spring return to original position upon de-energization
  • Compatible with SPST control devices
  • -40°F to 136°F (-40°C to 58°C) ambient temperature range
  • UL 873 listed
  • 5 to 95% RH non-condensing humidity tolerance
Catalog No. 50-276-2200 Supplier Grainger Supplier No. 161X20
This 240V electric actuator provides two-position operation for dampers, valves, and related equipment requiring a return to the normal position upon power interruption. The assembly features a rugged, die-cast aluminum enclosure designed for durability and is suitable for use in hazardous locations when configured appropriately. It operates with a two-wire control circuit and utilizes an oil-immersed motor and gear train for reliable performance.
  • 60 lb-in. rated torque
  • Includes auxiliary SPDT switch
  • Spring return to original position upon de-energization
  • Compatible with SPST control devices
  • -40°F to 136°F (-40°C to 58°C) ambient temperature range
  • UL 873 listed
  • 5 to 95% RH non-condensing humidity tolerance