The SmartX direct-coupled, spring-return actuator for proportional modulation control of dampers and valves in HVAC systems. This maintenance-free unit utilizes a microprocessor-controlled motor for precise operation and is designed for direct mounting to round or square shafts. Reliable in confined spaces, while a mechanical spring-return feature provides fail-safe operation and positive close-off.
- Proportional control with 2 to 10 Vdc or 4 to 20 mA signal support
- Mechanical clockwise or counterclockwise spring return for fail-safe operation
- NEMA 2 and IP54 rated housing for protection against dust and moisture
- Universal damper shaft clamp for round shafts up to 5/8 in. (15 mm) or 1/2 in. (13 mm) square
- Visual position indicator for simplified monitoring
- Adjustable rotation limiting from 40° to 95°
- Operating temperature range of -22°F to 140°F